Learn More
Grainger Threaded Shank Drill #30 HSS
Catalog No. 502764318
Encompass_Preferred
The Grainger 30-16W719 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw machine length drill point. It has a 118 degree point angle and is uncoated with an overall length of 1-1/2". The bit diameter is 1/8" and it has a bright finish. This threaded shank drill is designed for use in hand drills or drill presses on metal, plastic, and wood. It features a screw machine length drill point for accurate starting and clean holes in production drilling applications. The high-speed steel construction provides strength and heat resistance.
Catalog No. 50-276-4318 Supplier Grainger Supplier No. 16W719
The Grainger 30-16W719 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw machine length drill point. It has a 118 degree point angle and is uncoated with an overall length of 1-1/2". The bit diameter is 1/8" and it has a bright finish. This threaded shank drill is designed for use in hand drills or drill presses on metal, plastic, and wood. It features a screw machine length drill point for accurate starting and clean holes in production drilling applications. The high-speed steel construction provides strength and heat resistance.