Learn More
Grainger Threaded Shank Drill #40 Cobalt
Supplier:  Grainger 16W782
Encompass_Preferred
The Grainger 16W782 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw point tip. It has a length of 2-1/2" and is designed for use in metal, wood, and plastic. The bit features a 135 degree split point tip that starts drilling with less thrust and torque. It can be used in hand drills or drill presses. This USA-made drill bit is ideal for a variety of drilling applications.
Catalog No. 50-276-3676