Learn More
Grainger Threaded Shank Drill #40 Cobalt
Catalog No. 502763675
Encompass_Preferred
The Grainger 16W760 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw point tip. It has a length of 4" and is designed for use in metal, wood, and plastic. The bit features a 135 degree split point tip that starts drilling faster with less walking. It can be used in hand drills or drill presses and is made in the USA. The bit accepts a 1/4" hex quick change chuck for fast bit changes.
Catalog No. 50-276-3675 Supplier Grainger Supplier No. 16W760
The Grainger 16W760 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw point tip. It has a length of 4" and is designed for use in metal, wood, and plastic. The bit features a 135 degree split point tip that starts drilling faster with less walking. It can be used in hand drills or drill presses and is made in the USA. The bit accepts a 1/4" hex quick change chuck for fast bit changes.