Learn More
Perkin Elmer US LLC Threaded Spray Chamber Drain Line with PTFE Tubing for NexION 1000/2000
Catalog No. 502112191
Encompass_Preferred
The 32 cm PTFE drain line has a 1/4-28 spray chamber fitting. 160 cm of additional 0.8 mm I.D. drain line tubing is included.
Catalog No. 50-211-2191 Supplier Perkin Elmer US LLC Supplier No. N8152456
The 32 cm PTFE drain line has a 1/4-28 spray chamber fitting. 160 cm of additional 0.8 mm I.D. drain line tubing is included.