Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502767244
Encompass_Preferred
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter and 1/4" minimum hole size. It has a 60 degree threading angle and can cut unified coarse, unified fine, and metric threads. The overall length is 5" and it has a TiN coating for increased tool life. This threading tool is made in the USA by Micro 100 and can be used on lathes, screw machines, and CNC machines. The tool comes with a 30 day money back guarantee if not completely satisfied with performance.
Catalog No. 50-276-7244 Supplier Grainger Supplier No. 16T398
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter and 1/4" minimum hole size. It has a 60 degree threading angle and can cut unified coarse, unified fine, and metric threads. The overall length is 5" and it has a TiN coating for increased tool life. This threading tool is made in the USA by Micro 100 and can be used on lathes, screw machines, and CNC machines. The tool comes with a 30 day money back guarantee if not completely satisfied with performance.