Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502767568
Encompass_Preferred
The Grainger 16T233 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lowering. The hydraulic system is sealed and protected from dirt and moisture. This high quality bottle jack is designed for shop use and lifting equipment, dies, molds and more up to 32,000 pounds.
Catalog No. 50-276-7568 Supplier Grainger Supplier No. 16T233
The Grainger 16T233 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lowering. The hydraulic system is sealed and protected from dirt and moisture. This high quality bottle jack is designed for shop use and lifting equipment, dies, molds and more up to 32,000 pounds.