Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502768264
Encompass_Preferred
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 5" and it has a TiN coating for increased tool life. This threading tool is made in the USA by Micro 100 and can be used on lathes, screw machines, and CNC machines. The 1/2" shank allows it to be held securely in tool holders. It is a quality threading tool good for both production threading and general shop use.
Catalog No. 50-276-8264 Supplier Grainger Supplier No. 16T243
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 5" and it has a TiN coating for increased tool life. This threading tool is made in the USA by Micro 100 and can be used on lathes, screw machines, and CNC machines. The 1/2" shank allows it to be held securely in tool holders. It is a quality threading tool good for both production threading and general shop use.