Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502763857
Encompass_Preferred
The Micro 100 1/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 1/8" shank diameter and is designed for internal threading applications. The tool comes with one DCMT 060204 threading insert which provides a sharp cutting edge and positive rake angles for efficient chip removal. The insert is made from micro-grain carbide for wear resistance and durability. This threading tool allows quick insert changes and reduced downtime. It is well-suited for production threading applications in steels, cast iron, stainless steel, and hard materials. The tool provides accurate thread dimensions and surface finishes.
Catalog No. 50-276-3857 Supplier Grainger Supplier No. 16T060
The Micro 100 1/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 1/8" shank diameter and is designed for internal threading applications. The tool comes with one DCMT 060204 threading insert which provides a sharp cutting edge and positive rake angles for efficient chip removal. The insert is made from micro-grain carbide for wear resistance and durability. This threading tool allows quick insert changes and reduced downtime. It is well-suited for production threading applications in steels, cast iron, stainless steel, and hard materials. The tool provides accurate thread dimensions and surface finishes.