Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502763996
Encompass_Preferred
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter and an overall length of 5". It has a 60 degree threading point and can cut unified coarse, unified fine, and metric threads. The tool has an AlTiN coating for increased tool life and lubricity. It is made in the USA by Micro 100 and is well-suited for production threading applications on lathes, screw machines, and CNC lathes. The tool provides accurate threading with optimized chip flow and evacuation.
Catalog No. 50-276-3996 Supplier Grainger Supplier No. 16T070
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter and an overall length of 5". It has a 60 degree threading point and can cut unified coarse, unified fine, and metric threads. The tool has an AlTiN coating for increased tool life and lubricity. It is made in the USA by Micro 100 and is well-suited for production threading applications on lathes, screw machines, and CNC lathes. The tool provides accurate threading with optimized chip flow and evacuation.