Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502768406
Encompass_Preferred
The Micro 100 Threading Tool is a 1/2" shank threading tool with a 60 degree threading insert. It has a 0.062" minimum minor diameter and can cut up to 8 threads per inch. The tool body is made of C2 carbide and uses the TPG432 style threading insert, which is sold separately. The maximum insert size capacity is 5/16"x5/16". This is a high performance threading tool well-suited for production threading applications.
Catalog No. 50-276-8406 Supplier Grainger Supplier No. 16T241
The Micro 100 Threading Tool is a 1/2" shank threading tool with a 60 degree threading insert. It has a 0.062" minimum minor diameter and can cut up to 8 threads per inch. The tool body is made of C2 carbide and uses the TPG432 style threading insert, which is sold separately. The maximum insert size capacity is 5/16"x5/16". This is a high performance threading tool well-suited for production threading applications.