Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502767281
Encompass_Preferred
The Grainger website features internal threading taps used for creating internal threads in holes. They come in a variety of sizes, thread pitches, materials like high-speed steel and carbide, and coatings. Key features include the chamfered entering end for easy starting, and the tapered thread for easy insertion and alignment. They are available in plug, taper, and bottoming styles to suit different threading applications. Overall, these taps provide an efficient method of generating internal threads to precise specifications.
Catalog No. 50-276-7281 Supplier Grainger Supplier No. 16T395
The Grainger website features internal threading taps used for creating internal threads in holes. They come in a variety of sizes, thread pitches, materials like high-speed steel and carbide, and coatings. Key features include the chamfered entering end for easy starting, and the tapered thread for easy insertion and alignment. They are available in plug, taper, and bottoming styles to suit different threading applications. Overall, these taps provide an efficient method of generating internal threads to precise specifications.