Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502768372
Encompass_Preferred
The Micro 100 1/2" Shank Threading Tool is a high-speed steel threading tool with a 1/2" shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 5" and it has a 3/8" minor diameter. This threading tool is made in the USA out of C2 grade high-speed steel for good wear resistance. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, plastics, and more. The Micro 100 threading tool offers accurate single point threading in a durable tool.
Catalog No. 50-276-8372 Supplier Grainger Supplier No. 16T251
The Micro 100 1/2" Shank Threading Tool is a high-speed steel threading tool with a 1/2" shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 5" and it has a 3/8" minor diameter. This threading tool is made in the USA out of C2 grade high-speed steel for good wear resistance. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, plastics, and more. The Micro 100 threading tool offers accurate single point threading in a durable tool.