Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502765143
Encompass_Preferred
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and metric threads. The overall length is 5" and it has an American National thread size of 5/8"-11. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC lathes. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool offers accurate threading and extended tool life.
Catalog No. 50-276-5143 Supplier Grainger Supplier No. 16T131
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and metric threads. The overall length is 5" and it has an American National thread size of 5/8"-11. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC lathes. It features a TiN coating for increased tool life and lubricity. The Micro 100 threading tool offers accurate threading and extended tool life.