Learn More
Grainger Threading Tool 1/2 Carbide
Supplier:  Grainger 16T237
Encompass_Preferred
The Micro 100 Threading Tool is a 1/2" shank threading tool with a 60 degree included angle and a 0.040" nose radius. It has a TiN coating and is made of C2 micrograin carbide for wear resistance and strength. The tool has an American National thread form and can cut unified coarse, unified fine, and 8 pitch threads. It has coolant through the center to improve tool life. The threading tool has a screw lock style holder and is made in the USA. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The tool provides accurate threading with optimized chip control.
Catalog No. 50-276-8468