Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502765599
Encompass_Preferred
The Grainger 16T436 is a 16-ton hydraulic bottle jack made of steel with a lifting range of 10-15/16 inches to 17-1/8 inches. It has a built-in overload valve and bypass device and meets ASME PALD standards. The jack has a saddle diameter of 5-1/8 inches and an extended height of 17-1/8 inches. It is designed for garage services, farm equipment repair, and industrial maintenance applications. The 16T436 has a 12 month warranty and weighs 43 pounds.
Catalog No. 50-276-5599 Supplier Grainger Supplier No. 16T436
The Grainger 16T436 is a 16-ton hydraulic bottle jack made of steel with a lifting range of 10-15/16 inches to 17-1/8 inches. It has a built-in overload valve and bypass device and meets ASME PALD standards. The jack has a saddle diameter of 5-1/8 inches and an extended height of 17-1/8 inches. It is designed for garage services, farm equipment repair, and industrial maintenance applications. The 16T436 has a 12 month warranty and weighs 43 pounds.