Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502763960
Encompass_Preferred
The Grainger 16T052 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This compact, lightweight jack has a welded base for stability and swivel saddle to adjust to any angle. It is designed for portability and ease of use with a built-in handle and two wheels. The 16T052 hydraulic bottle jack is ideal for lifting equipment, vehicles, and machinery up to 32,000 pounds. It requires hydraulic fluid, sold separately. This is a high-quality jack made in the USA.
Catalog No. 50-276-3960 Supplier Grainger Supplier No. 16T052
The Grainger 16T052 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This compact, lightweight jack has a welded base for stability and swivel saddle to adjust to any angle. It is designed for portability and ease of use with a built-in handle and two wheels. The 16T052 hydraulic bottle jack is ideal for lifting equipment, vehicles, and machinery up to 32,000 pounds. It requires hydraulic fluid, sold separately. This is a high-quality jack made in the USA.