Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502763962
Encompass_Preferred
The Micro 100 1/2" Carbide Threading Tool is a high-performance threading tool designed for cutting a variety of materials including steel, stainless steel, cast iron, aluminum, and more. It features a durable carbide cutting edge and can cut 16 threads per inch. The tool has a 1/2" shank diameter and an overall length of 5". It can be used on lathes, screw machines, and CNC equipment. The Micro 100 threading tool delivers accurate, chatter-free threading and long tool life.
Catalog No. 50-276-3962 Supplier Grainger Supplier No. 16T065
The Micro 100 1/2" Carbide Threading Tool is a high-performance threading tool designed for cutting a variety of materials including steel, stainless steel, cast iron, aluminum, and more. It features a durable carbide cutting edge and can cut 16 threads per inch. The tool has a 1/2" shank diameter and an overall length of 5". It can be used on lathes, screw machines, and CNC equipment. The Micro 100 threading tool delivers accurate, chatter-free threading and long tool life.