Learn More
Grainger Threading Tool 1/2 Carbide
Catalog No. 502763986
Encompass_Preferred
The Grainger 16T059 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing Electric Motor. It has a 1.15 service factor, CCW rotation when facing shaft end, and features a cast iron frame, ball bearing construction, and Class F insulation. This motor is suitable for industrial applications requiring reliable performance. With a full load current of 12.2 Amps, it can provide 0.56 kW of mechanical power from electrical input. The motor dimensions are 9-1/4" long by 7-3/16" wide by 8-1/4" high and it weighs 38 lbs.
Catalog No. 50-276-3986 Supplier Grainger Supplier No. 16T059
The Grainger 16T059 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing Electric Motor. It has a 1.15 service factor, CCW rotation when facing shaft end, and features a cast iron frame, ball bearing construction, and Class F insulation. This motor is suitable for industrial applications requiring reliable performance. With a full load current of 12.2 Amps, it can provide 0.56 kW of mechanical power from electrical input. The motor dimensions are 9-1/4" long by 7-3/16" wide by 8-1/4" high and it weighs 38 lbs.