Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502766431
Encompass_Preferred
The Micro 100 Threading Tool is a 1/4" shank threading tool with a 60 degree threading insert. It has a 0.040" minimum minor diameter and can cut up to 8 threads per inch. The insert is made of C2 grade carbide and allows high speed threading in most materials. The tool has a TiN coating for increased tool life. It can be used for both internal and external threading applications. The shank allows it to fit standard tool holders. This Micro 100 threading tool is well-suited for light duty threading operations.
Catalog No. 50-276-6431 Supplier Grainger Supplier No. 16T347
The Micro 100 Threading Tool is a 1/4" shank threading tool with a 60 degree threading insert. It has a 0.040" minimum minor diameter and can cut up to 8 threads per inch. The insert is made of C2 grade carbide and allows high speed threading in most materials. The tool has a TiN coating for increased tool life. It can be used for both internal and external threading applications. The shank allows it to fit standard tool holders. This Micro 100 threading tool is well-suited for light duty threading operations.