Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502767320
Encompass_Preferred
The Grainger 16T108 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-1/8 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and protected from dirt and moisture. This heavy duty jack is designed for shop use and farm equipment lifting. With a 16 ton capacity, it can safely lift up to 32,000 pounds. The jack weighs 43 pounds and measures 7 x 11 x 21 inches. It is made in the USA and comes with a one year warranty from Hein-Werner.
Catalog No. 50-276-7320 Supplier Grainger Supplier No. 16T108
The Grainger 16T108 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-1/8 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and protected from dirt and moisture. This heavy duty jack is designed for shop use and farm equipment lifting. With a 16 ton capacity, it can safely lift up to 32,000 pounds. The jack weighs 43 pounds and measures 7 x 11 x 21 inches. It is made in the USA and comes with a one year warranty from Hein-Werner.