Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502767396
Encompass_Preferred
The Micro 100 Threading Tool is a high-performance threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It features a TiN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its precision ground thread profile, it is capable of producing accurate internal and external threads in blind holes and through holes. The tool has an included angle of 60 degrees and is made of micro-grain carbide for strength and durability. Its high helix angle and polished flutes allow efficient chip evacuation while threading.
Catalog No. 50-276-7396 Supplier Grainger Supplier No. 16T164
The Micro 100 Threading Tool is a high-performance threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It features a TiN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its precision ground thread profile, it is capable of producing accurate internal and external threads in blind holes and through holes. The tool has an included angle of 60 degrees and is made of micro-grain carbide for strength and durability. Its high helix angle and polished flutes allow efficient chip evacuation while threading.