Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502767569
Encompass_Preferred
The Grainger 16T250 is a 16-tooth, 250mm diameter circular saw blade with a 20mm arbor hole and a 2.8mm kerf width. It is designed for cutting wood, plywood, and chipboard with a maximum depth of cut at 90° of 66mm. This carbide-tipped blade has a thin kerf for fast, smooth cuts and features laser cut stabilizer vents to reduce vibration. With a 5 degree positive hook angle, this saw blade is optimized for cross-cutting and ripping solid wood stock. It should be run at a speed of 4800 RPM with a negative 5 degree rake angle when cutting. This durable saw blade is ideal for precise, clean cuts in woodworking applications.
Catalog No. 50-276-7569 Supplier Grainger Supplier No. 16T250
The Grainger 16T250 is a 16-tooth, 250mm diameter circular saw blade with a 20mm arbor hole and a 2.8mm kerf width. It is designed for cutting wood, plywood, and chipboard with a maximum depth of cut at 90° of 66mm. This carbide-tipped blade has a thin kerf for fast, smooth cuts and features laser cut stabilizer vents to reduce vibration. With a 5 degree positive hook angle, this saw blade is optimized for cross-cutting and ripping solid wood stock. It should be run at a speed of 4800 RPM with a negative 5 degree rake angle when cutting. This durable saw blade is ideal for precise, clean cuts in woodworking applications.