Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502767586
Encompass_Preferred
The Grainger 16T343 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and operates on 60 Hz power. The motor is thermally protected and rated for continuous duty applications. It can provide 0.56 kW of output power. The 16T343 has a NEMA standard 56C frame size with standard starting torque and breakdown torque. This electric motor is suitable for various industrial applications requiring up to 3/4 horsepower.
Catalog No. 50-276-7586 Supplier Grainger Supplier No. 16T343
The Grainger 16T343 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and operates on 60 Hz power. The motor is thermally protected and rated for continuous duty applications. It can provide 0.56 kW of output power. The 16T343 has a NEMA standard 56C frame size with standard starting torque and breakdown torque. This electric motor is suitable for various industrial applications requiring up to 3/4 horsepower.