Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502766395
Encompass_Preferred
The Micro 100 Threading Tool is a 1/4 inch shank threading tool with a 60 degree threading insert. It has a 0.040 inch nose radius and can cut unified thread sizes from 4-56 to 1/4-20 as well as metric thread sizes from M1.6 to M6. The tool holder accepts DCMT/CCMT style inserts and has an overall length of 4 inches. The threading tool features precision ground carbide construction for strength and tool life. It's designed for internal and external threading operations on CNC lathes.
Catalog No. 50-276-6395 Supplier Grainger Supplier No. 16T352
The Micro 100 Threading Tool is a 1/4 inch shank threading tool with a 60 degree threading insert. It has a 0.040 inch nose radius and can cut unified thread sizes from 4-56 to 1/4-20 as well as metric thread sizes from M1.6 to M6. The tool holder accepts DCMT/CCMT style inserts and has an overall length of 4 inches. The threading tool features precision ground carbide construction for strength and tool life. It's designed for internal and external threading operations on CNC lathes.