Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502766401
Encompass_Preferred
The Grainger 1/4" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree included angle and 16 threads per inch. The overall length is 3-1/2" and it has a 1/4" shank diameter to fit standard tool holders. Made in the USA, this durable threading tool can cut accurate internal and external unified threads in manual or CNC machine shop applications.
Catalog No. 50-276-6401 Supplier Grainger Supplier No. 16T338
The Grainger 1/4" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree included angle and 16 threads per inch. The overall length is 3-1/2" and it has a 1/4" shank diameter to fit standard tool holders. Made in the USA, this durable threading tool can cut accurate internal and external unified threads in manual or CNC machine shop applications.