Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502766920
Encompass_Preferred
The Grainger 16T349 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This hydraulic jack has a two-piece handle for easy pumping and is designed for use on hard level surfaces. It has a welded base for stability with rubber saddle to prevent slippage. The 16T349 bottle jack has a maximum lift height of 16-1/4 inches and conforms to ASME PALD standards. This heavy duty hydraulic jack is recommended for use in auto repair shops and construction sites for lifting equipment, vehicles, and machinery.
Catalog No. 50-276-6920 Supplier Grainger Supplier No. 16T349
The Grainger 16T349 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This hydraulic jack has a two-piece handle for easy pumping and is designed for use on hard level surfaces. It has a welded base for stability with rubber saddle to prevent slippage. The 16T349 bottle jack has a maximum lift height of 16-1/4 inches and conforms to ASME PALD standards. This heavy duty hydraulic jack is recommended for use in auto repair shops and construction sites for lifting equipment, vehicles, and machinery.