Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502767347
Encompass_Preferred
The Micro 100 1/4" Shank Threading Tool is made of C2 grade high speed steel, offering good wear resistance. It features a 60-degree threading point and can cut unified coarse, unified fine, and metric threads. With a 1/4" shank diameter and 3-1/2" overall length, it fits tool holders for lathes, screw machines, and CNC machines. Suitable for machining steel, stainless steel, cast iron, aluminum, and plastics, this USA-made tool is ideal for production and general workshop threading.
Catalog No. 50-276-7347 Supplier Grainger Supplier No. 16T165
The Micro 100 1/4" Shank Threading Tool is made of C2 grade high speed steel, offering good wear resistance. It features a 60-degree threading point and can cut unified coarse, unified fine, and metric threads. With a 1/4" shank diameter and 3-1/2" overall length, it fits tool holders for lathes, screw machines, and CNC machines. Suitable for machining steel, stainless steel, cast iron, aluminum, and plastics, this USA-made tool is ideal for production and general workshop threading.