Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502767375
Encompass_Preferred
The Grainger 16T166 is a 16-ton hydraulic bottle jack made of steel with a lifting range of 10-15/16 inches to 16-1/2 inches. It has a built-in overload valve and bypass device and meets ASME PALD standards. The jack has a saddle diameter of 5-1/8 inches and an extended height of 21-7/8 inches. It is designed for garage, auto repair shop, and industrial plant use. The 16T166 hydraulic jack has a 1-year warranty from Grainger against defects in materials and workmanship.
Catalog No. 50-276-7375 Supplier Grainger Supplier No. 16T166
The Grainger 16T166 is a 16-ton hydraulic bottle jack made of steel with a lifting range of 10-15/16 inches to 16-1/2 inches. It has a built-in overload valve and bypass device and meets ASME PALD standards. The jack has a saddle diameter of 5-1/8 inches and an extended height of 21-7/8 inches. It is designed for garage, auto repair shop, and industrial plant use. The 16T166 hydraulic jack has a 1-year warranty from Grainger against defects in materials and workmanship.