Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502765326
Encompass_Preferred
The Grainger 16T342 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor, thermally protected, manual reset overload protector, and meets NEMA design B standards. This motor can be used for various industrial applications requiring 3/4 HP.
Catalog No. 50-276-5326 Supplier Grainger Supplier No. 16T342
The Grainger 16T342 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 1.15 service factor, thermally protected, manual reset overload protector, and meets NEMA design B standards. This motor can be used for various industrial applications requiring 3/4 HP.