Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502766093
Encompass_Preferred
The Grainger 1/4" Carbide Threading Tool is a high-speed steel threading insert designed for cutting threads in steel, stainless steel, cast iron, aluminum, and other metals. It features a TiN coating for wear resistance and lubricity. The insert has a 16 mm shank diameter and fits standard tool holders. It can cut 60 degree threads from 4-56 to 1/4-20 TPI in a variety of materials. The carbide composition provides strength and heat resistance for optimal thread cutting performance. This durable threading tool is made in the USA and ideal for production threading applications.
Catalog No. 50-276-6093 Supplier Grainger Supplier No. 16T254
The Grainger 1/4" Carbide Threading Tool is a high-speed steel threading insert designed for cutting threads in steel, stainless steel, cast iron, aluminum, and other metals. It features a TiN coating for wear resistance and lubricity. The insert has a 16 mm shank diameter and fits standard tool holders. It can cut 60 degree threads from 4-56 to 1/4-20 TPI in a variety of materials. The carbide composition provides strength and heat resistance for optimal thread cutting performance. This durable threading tool is made in the USA and ideal for production threading applications.