Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502767052
Encompass_Preferred
The Grainger 16T107 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-15 inches and a lowered height of 9.5 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two-piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and has an overload valve for safety. This heavy duty jack is designed for shop use and farm equipment lifting. With its sturdy steel construction, it provides portable power for jobs up to 16 tons.
Catalog No. 50-276-7052 Supplier Grainger Supplier No. 16T107
The Grainger 16T107 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-15 inches and a lowered height of 9.5 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two-piece handle for easy pumping and controlled lifting. The hydraulic system is sealed and has an overload valve for safety. This heavy duty jack is designed for shop use and farm equipment lifting. With its sturdy steel construction, it provides portable power for jobs up to 16 tons.