Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502768344
Encompass_Preferred
The Grainger 16T431 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This hydraulic jack has a two-piece handle for easy pumping and is designed for use on hard level surfaces. It has a welded base for stability with rubber saddle to prevent slippage. The 16T431 bottle jack has a maximum lift height of 16-1/4 inches and conforms to ASME PALD standards. This heavy duty hydraulic jack is rated for 16 tons lifting capacity.
Catalog No. 50-276-8344 Supplier Grainger Supplier No. 16T431
The Grainger 16T431 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This hydraulic jack has a two-piece handle for easy pumping and is designed for use on hard level surfaces. It has a welded base for stability with rubber saddle to prevent slippage. The 16T431 bottle jack has a maximum lift height of 16-1/4 inches and conforms to ASME PALD standards. This heavy duty hydraulic jack is rated for 16 tons lifting capacity.