Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502764951
Encompass_Preferred
The Grainger 16T350 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings, manual reset thermal overload protection, and meets NEMA design standards. This 1/3 HP motor can provide 0.25 kW of output power.
Catalog No. 50-276-4951 Supplier Grainger Supplier No. 16T350
The Grainger 16T350 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings, manual reset thermal overload protection, and meets NEMA design standards. This 1/3 HP motor can provide 0.25 kW of output power.