Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502768478
Encompass_Preferred
The Grainger 16T337 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-woven fibers in a medium grade for cleaning and scouring. This 6 inch by 9 inch hand pad has an open lapping that exposes the fibers for scrubbing and is ideal for cleaning pots, pans, dishes, flatware, sinks, showers, tubs, fiberglass, porcelain, stainless steel, chrome, glass, plastics, enamel, and more. It effectively removes grease, grime, food residue, and dirt without scratching. The pad can be used wet or dry.
Catalog No. 50-276-8478 Supplier Grainger Supplier No. 16T337
The Grainger 16T337 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-woven fibers in a medium grade for cleaning and scouring. This 6 inch by 9 inch hand pad has an open lapping that exposes the fibers for scrubbing and is ideal for cleaning pots, pans, dishes, flatware, sinks, showers, tubs, fiberglass, porcelain, stainless steel, chrome, glass, plastics, enamel, and more. It effectively removes grease, grime, food residue, and dirt without scratching. The pad can be used wet or dry.