Learn More
Grainger Threading Tool 1/4 Carbide
Catalog No. 502765596
Encompass_Preferred
The Grainger 16T430 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-15 inches and a lowered height of 9.5 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for use on hard level surfaces and has a built-in safety valve and bypass system. The jack meets ASME PALD standards and has a 1 year warranty. With a lifting capacity of 32,000 lbs, this heavy duty hydraulic jack is suitable for automotive and industrial lifting applications.
Catalog No. 50-276-5596 Supplier Grainger Supplier No. 16T430
The Grainger 16T430 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-15 inches and a lowered height of 9.5 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for use on hard level surfaces and has a built-in safety valve and bypass system. The jack meets ASME PALD standards and has a 1 year warranty. With a lifting capacity of 32,000 lbs, this heavy duty hydraulic jack is suitable for automotive and industrial lifting applications.