Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502763987
Encompass_Preferred
The Grainger 16T077 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The jack's capacity is 16 tons (32,000 lbs). It weighs 43 pounds and has a one year warranty. The jack is designed for use on hard level surfaces for lifting equipment, dies, molds, and more. It meets ASME PALD standards.
Catalog No. 50-276-3987 Supplier Grainger Supplier No. 16T077
The Grainger 16T077 is a 16 ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It has a two piece handle for easy pumping and controlled lifting. The jack's capacity is 16 tons (32,000 lbs). It weighs 43 pounds and has a one year warranty. The jack is designed for use on hard level surfaces for lifting equipment, dies, molds, and more. It meets ASME PALD standards.