Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502764091
Encompass_Preferred
The Grainger 16T074 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield. It can provide 0.56 kW of output power. The 16T074 would be a good choice for powering pumps, fans, conveyors, compressors, machine tools, or other industrial equipment needing up to 3/4 horsepower.
Catalog No. 50-276-4091 Supplier Grainger Supplier No. 16T074
The Grainger 16T074 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield. It can provide 0.56 kW of output power. The 16T074 would be a good choice for powering pumps, fans, conveyors, compressors, machine tools, or other industrial equipment needing up to 3/4 horsepower.