Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502765982
Encompass_Preferred
The Micro 100 1/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 1/8" shank diameter and can cut unified thread sizes from 4-56 to 1/4-20 as well as metric thread sizes from M1.6 to M6. The tool comes with one DCMT 070204 threading insert and can cut a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The threading tool has a TiN coating for increased tool life.
Catalog No. 50-276-5982 Supplier Grainger Supplier No. 16T139
The Micro 100 1/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 1/8" shank diameter and can cut unified thread sizes from 4-56 to 1/4-20 as well as metric thread sizes from M1.6 to M6. The tool comes with one DCMT 070204 threading insert and can cut a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The threading tool has a TiN coating for increased tool life.