Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502768521
Encompass_Preferred
The Micro 100 1/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree included angle and a 1/8" shank diameter. It has a right hand thread direction and can cut unified thread sizes from 4-56 to 1/4"-20. The overall length is 3-1/2" and it has a TiN coating for increased tool life and lubricity. This threading tool is made in the USA by Micro 100 and is well-suited for production threading applications where tool life and performance are important.
Catalog No. 50-276-8521 Supplier Grainger Supplier No. 16T147
The Micro 100 1/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree included angle and a 1/8" shank diameter. It has a right hand thread direction and can cut unified thread sizes from 4-56 to 1/4"-20. The overall length is 3-1/2" and it has a TiN coating for increased tool life and lubricity. This threading tool is made in the USA by Micro 100 and is well-suited for production threading applications where tool life and performance are important.