Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502765178
Encompass_Preferred
The Micro 100 Threading Tool is a high-speed steel threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It features a 60 degree threading insert with TiN coating for wear resistance when cutting a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The tool provides accurate internal and external 60 degree threads.
Catalog No. 50-276-5178 Supplier Grainger Supplier No. 16T148
The Micro 100 Threading Tool is a high-speed steel threading tool with an 8mm shank diameter and 16mm maximum cutting diameter. It features a 60 degree threading insert with TiN coating for wear resistance when cutting a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The tool provides accurate internal and external 60 degree threads.