Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502768378
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 3 inches and it has an American National Standard stub length of 2 inches. This durable tool is made in the USA and well-suited for production threading applications. The Micro 100 brand is known for quality and performance.
Catalog No. 50-276-8378 Supplier Grainger Supplier No. 16T140
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter. It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The overall length is 3 inches and it has an American National Standard stub length of 2 inches. This durable tool is made in the USA and well-suited for production threading applications. The Micro 100 brand is known for quality and performance.