Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502763954
Encompass_Preferred
The Grainger 1/8" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree included angle and 16 threads per inch. The overall length is 3-1/2" and it has a square shank that fits 3/8" tool holders. This durable threading tool provides accurate threading and long tool life. It is made in the USA.
Catalog No. 50-276-3954 Supplier Grainger Supplier No. 16T080
The Grainger 1/8" Carbide Threading Tool is a high-speed steel threading tool with a carbide tip designed for cutting threads in steel, cast iron, aluminum, and other metals. It features a 60 degree included angle and 16 threads per inch. The overall length is 3-1/2" and it has a square shank that fits 3/8" tool holders. This durable threading tool provides accurate threading and long tool life. It is made in the USA.