Learn More
Grainger Threading Tool 1/8 Carbide
Catalog No. 502763997
Encompass_Preferred
The Grainger 1/8" Carbide Threading Tool is a high-speed steel threading tool designed for cutting internal and external right-hand threads. It features a 60 degree threading point with a 1/8" shank diameter and an overall length of 5". The carbide tip provides durability, heat resistance, and a sharp cutting edge for clean thread cuts in steel, cast iron, aluminum, and other metals. This single-point threading tool allows both axial and radial depth of cut adjustments for precision thread forming. Its compact size makes it well-suited for threading in tight spaces.
Catalog No. 50-276-3997 Supplier Grainger Supplier No. 16T072
The Grainger 1/8" Carbide Threading Tool is a high-speed steel threading tool designed for cutting internal and external right-hand threads. It features a 60 degree threading point with a 1/8" shank diameter and an overall length of 5". The carbide tip provides durability, heat resistance, and a sharp cutting edge for clean thread cuts in steel, cast iron, aluminum, and other metals. This single-point threading tool allows both axial and radial depth of cut adjustments for precision thread forming. Its compact size makes it well-suited for threading in tight spaces.