Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502763908
Encompass_Preferred
The Micro 100 3/16" Threading Tool is a high speed steel threading tool with a 60 degree included angle and a 16 TPI thread. It has a 3/16" shank diameter and an overall length of 3-1/2". The insert is replaceable with additional inserts available. This single point threading tool can be used for both internal and external threading operations. The tool comes with a TiN coating for increased tool life and lubricity. It is made in the USA by Micro 100, a manufacturer specializing in precision cutting tools. The tool is well-suited for production threading applications.
Catalog No. 50-276-3908 Supplier Grainger Supplier No. 16T095
The Micro 100 3/16" Threading Tool is a high speed steel threading tool with a 60 degree included angle and a 16 TPI thread. It has a 3/16" shank diameter and an overall length of 3-1/2". The insert is replaceable with additional inserts available. This single point threading tool can be used for both internal and external threading operations. The tool comes with a TiN coating for increased tool life and lubricity. It is made in the USA by Micro 100, a manufacturer specializing in precision cutting tools. The tool is well-suited for production threading applications.