Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765337
Encompass_Preferred
The Micro 100 3/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 3/8" shank diameter and is 5-1/2" overall length. The insert has a .040" nose radius and can cut unified thread sizes from 4-56 to 1"-8. It has an American National thread form and features TiN coating for increased tool life. The tool comes with one DGT060204 threading insert. It is made in the USA.
Catalog No. 50-276-5337 Supplier Grainger Supplier No. 16T317
The Micro 100 3/8" Shank Threading Tool is a high speed steel threading tool with a 60 degree threading insert. It has a 3/8" shank diameter and is 5-1/2" overall length. The insert has a .040" nose radius and can cut unified thread sizes from 4-56 to 1"-8. It has an American National thread form and features TiN coating for increased tool life. The tool comes with one DGT060204 threading insert. It is made in the USA.