Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502765965
Encompass_Preferred
The Grainger 16T150 is a 16-ton hydraulic bottle jack made of steel with a lifting range of 10-15 inches. It has a built-in overload valve and bypass device for safety and meets ASME PALD standards. This compact, portable jack has a 7-1/8 inch ram diameter and measures 14-1/2 inches high when collapsed. It weighs 43 pounds and has a handle for carrying and positioning. The 16T150 bottle jack is designed for shop use and general maintenance applications.
Catalog No. 50-276-5965 Supplier Grainger Supplier No. 16T150
The Grainger 16T150 is a 16-ton hydraulic bottle jack made of steel with a lifting range of 10-15 inches. It has a built-in overload valve and bypass device for safety and meets ASME PALD standards. This compact, portable jack has a 7-1/8 inch ram diameter and measures 14-1/2 inches high when collapsed. It weighs 43 pounds and has a handle for carrying and positioning. The 16T150 bottle jack is designed for shop use and general maintenance applications.