Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502766244
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16ER ANSI standard thread. It has a 60 degree included angle and can cut unified screw threads from 4-48 to 1"-8. The overall length is 5" and it weighs 0.19 lbs. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features a TiN coating for increased tool life. The Micro 100 threading tool is made in the USA.
Catalog No. 50-276-6244 Supplier Grainger Supplier No. 16T271
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/8" shank diameter and 16ER ANSI standard thread. It has a 60 degree included angle and can cut unified screw threads from 4-48 to 1"-8. The overall length is 5" and it weighs 0.19 lbs. This single point threading tool allows quick threading passes on lathes, screw machines, and CNC machines. It features a TiN coating for increased tool life. The Micro 100 threading tool is made in the USA.