Learn More
Grainger Threading Tool 3/16 Carbide
Supplier:  Grainger 16T102
Encompass_Preferred
The Grainger internal threading tools have a 3/16 inch shank diameter and are made of high speed steel. They feature a 60 degree threading angle and are available in multiple sizes including #4-40, #6-32, #8-32, #10-24, #10-32, and 1/4-20. The tools have an overall length of 3 to 4 inches depending on the thread size. They can be used for hand threading operations on lathes, screw machines, and machining centers. The high speed steel construction provides good wear resistance and cutting ability.
Catalog No. 50-276-4090