Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502766535
Encompass_Preferred
The Grainger 16T281 is a 16-ton bottle jack made of steel with a lifting range of 10-15/16 inches to 17-1/8 inches. It has a built-in overload valve and bypass device for controlled lifting. The jack's base is 7-3/4 x 5-1/8 inches and it stands 23-1/2 inches tall. It conforms to ASME PALD standards with a 150% factory test of 24 tons. This heavy duty hydraulic jack is designed for shop use and garage automotive work.
Catalog No. 50-276-6535 Supplier Grainger Supplier No. 16T281
The Grainger 16T281 is a 16-ton bottle jack made of steel with a lifting range of 10-15/16 inches to 17-1/8 inches. It has a built-in overload valve and bypass device for controlled lifting. The jack's base is 7-3/4 x 5-1/8 inches and it stands 23-1/2 inches tall. It conforms to ASME PALD standards with a 150% factory test of 24 tons. This heavy duty hydraulic jack is designed for shop use and garage automotive work.