Learn More
Grainger Threading Tool 3/16 Carbide
Catalog No. 502767126
Encompass_Preferred
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree threading point. It has an overall length of 4 inches and can cut unified coarse right hand threads from 4 to 56 threads per inch. The tool has an AlTiN nano coating for increased tool life and can machine a variety of materials including carbon steel, alloy steel, stainless steel, cast iron, aluminum, brass, bronze and plastics. It is made in the USA by Micro 100 and can be used on lathes, screw machines or CNC machines. The tool comes with a 30 day money back guarantee.
Catalog No. 50-276-7126 Supplier Grainger Supplier No. 16T298
The Micro 100 Threading Tool is a high speed steel threading tool with a 3/16 inch shank diameter and 60 degree threading point. It has an overall length of 4 inches and can cut unified coarse right hand threads from 4 to 56 threads per inch. The tool has an AlTiN nano coating for increased tool life and can machine a variety of materials including carbon steel, alloy steel, stainless steel, cast iron, aluminum, brass, bronze and plastics. It is made in the USA by Micro 100 and can be used on lathes, screw machines or CNC machines. The tool comes with a 30 day money back guarantee.